SY3371478175 Synthonix Corporation

Imidazo[1,2-a]pyridin-3-ylmethanol hydrochloride

  • MDL number MFCD13176617
  • Cas Number : 30489-62-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/3369.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/3369.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart