SY3371478180 Synthonix Corporation

3-Iodo-1-methyl-1H-indazole-5-carboxylic acid ethyl ester

  • MDL number MFCD11109380
  • Cas Number : 1092351-68-2
  • Molecular Weight : 330.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/3452.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/3452.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart