SY3371478228 Synthonix Corporation

Ethyl 1H-indazole-7-carboxylate

  • Cas Number : 885278-74-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link https://synthonix.com/6105.html

Safety Information

MSDS https://synthonix.com/6105.html
Availability details for
Quantity
  • Availability
  • Add to Cart