SY3371478273 Synthonix Corporation

(S)-3-(Methylsulfonyl)pyrrolidine

  • MDL number MFCD16294584
  • Cas Number : 290328-57-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin USA
Product Link https://synthonix.com/0288.html

Safety Information

MSDS https://synthonix.com/0288.html
hstatements H301 , H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart