SY3371478339 Synthonix Corporation

1-Methyl-1H-imidazole-2-amidine hydrochloride

  • MDL number MFCD09952683
  • Cas Number : 849600-64-2
  • Molecular Weight : 160.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/0674.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/0674.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart