SY3371478411 Synthonix Corporation

Methyl 5-aminopyrimidine-2-carboxylate

  • MDL number MFCD08234876
  • Cas Number : 73418-88-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/1663.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/1663.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart