SY3371478413 Synthonix Corporation

Methyl 2-hydroxypyridine-4-carboxylate

  • MDL number MFCD09258780
  • Cas Number : 89937-77-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/1682.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/1682.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart