SY3371478427 Synthonix Corporation

Methyl 1-diphenylmethyl-azetidine-3-carboxylate

  • MDL number MFCD03407521
  • Cas Number : 53871-06-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/1904.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/1904.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart