SY3371478437 Synthonix Corporation

Methyl 2-bromothiazole-4-carboxylate

  • MDL number MFCD06659908
  • Cas Number : 170235-26-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link https://synthonix.com/2154.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/2154.html
hstatements H302 , H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart