SY3371478446 Synthonix Corporation

2-Methoxy-4-(tributylstannyl)thiazole

  • MDL number MFCD07787372
  • Cas Number : 927391-09-1
  • Molecular Weight : 404.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Referigrator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/2257.html

Safety Information

signalword Danger
pcodes P261, P262, P264, P270, P273, P280
MSDS https://synthonix.com/2257.html
pictograms GHS06, GHS08, GHS09
hstatements H301, H312, H315, H319, H332, H410
Availability details for
Quantity
  • Availability
  • Add to Cart