SY3371478472 Synthonix Corporation

2-Methoxythiazole

  • MDL number MFCD01631143
  • Cas Number : 14542-13-3
  • Molecular Weight : 115.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/2608.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/2608.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart