SY3371478499 Synthonix Corporation

2-Methylpropanethioamide

  • MDL number MFCD07369538
  • Cas Number : 13515-65-6
  • Molecular Weight : 103.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/3101.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/3101.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart