SY3371478515 Synthonix Corporation

Methyl 4-chloro-7-azaindole-2-carboxylate

  • MDL number MFCD09839319
  • Cas Number : 871583-23-2
  • Molecular Weight : 210.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/3345.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/3345.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart