SY3371478519 Synthonix Corporation

7-Methylimidazo[1,2-a]pyridine-2-carboxylic acid ethyl ester

  • MDL number MFCD02187526
  • Cas Number : 70705-33-8
  • Molecular Weight : 204.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/3390.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/3390.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart