SY3371478596 Synthonix Corporation

Methyl 5,7-dichloropyrazolo[1,5-a]pyrimidine-3-carboxylate

  • Cas Number : 1053656-37-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/4255.html

Safety Information

MSDS https://synthonix.com/4255.html
Availability details for
Quantity
  • Availability
  • Add to Cart