SY3371478629 Synthonix Corporation

Methyl 8-Bromo-6-methylimidazo[1,2-a]pyridine-2-carboxylate

  • MDL number MFCD12026326
  • Cas Number : 1171424-92-2
  • Molecular Weight : 269.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/5083.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/5083.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart