SY3371478635 Synthonix Corporation

Methyl 5-chloro-1H-pyrrolo[2,3-b]pyridine-2-carboxylate

  • Cas Number : 952182-19-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/5235.html

Safety Information

MSDS https://synthonix.com/5235.html
Availability details for
Quantity
  • Availability
  • Add to Cart