SY3371478639 Synthonix Corporation

2-Methylthieno[3,2-c]pyridin-4(5H)-one

  • Cas Number : 59207-23-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/5279.html

Safety Information

MSDS https://synthonix.com/5279.html
Availability details for
Quantity
  • Availability
  • Add to Cart