SY3371478640 Synthonix Corporation

Methyl isoindoline-4-carboxylate hydrochloride

  • MDL number MFCD10700133
  • Cas Number : 127168-90-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/5356.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/5356.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart