SY3371478677 Synthonix Corporation

2-Methoxy-2-methylpropylamine

  • MDL number MFCD01821229
  • Cas Number : 89282-70-2
  • Molecular Weight : 103.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/6043.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/6043.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart