SY3371478683 Synthonix Corporation

5-(Methoxymethyl)-3-methylisoxazole-4-carboxylic acid

  • MDL number MFCD18839245
  • Cas Number : 1108712-47-5
  • Molecular Weight : 171.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/6216.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/6216.html
pictograms GHS07
hstatements H315, H317, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart