SY3371478722 Synthonix Corporation

N-(5-Methylthiazol-2-yl)-5,6,7,8-tetrahydro-4H-cyclohepta[b]thiophene-2-carboxamide

  • MDL number MFCD09431082
  • Cas Number : 925606-29-7
  • Molecular Weight : 292.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/6985.html

Safety Information

signalword Warning
pcodes P280
MSDS https://synthonix.com/6985.html
pictograms GHS06, GHS07
hstatements H301, H319
Availability details for
Quantity
  • Availability
  • Add to Cart