SY3371478748 Synthonix Corporation

3-(Nitromethyl)oxetane

  • MDL number MFCD12547196
  • Cas Number : 1313739-08-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/6280.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/6280.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart