SY3371478792 Synthonix Corporation

N-(3-Oxobutyl)phthalimide

  • MDL number MFCD00458653
  • Cas Number : 3783-77-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link https://synthonix.com/6017.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/6017.html
hstatements H302 , H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart