SY3371478802 Synthonix Corporation

2-Aminomethylpyrimidine

  • MDL number MFCD06212857
  • Cas Number : 75985-45-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/0030.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/0030.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart