SY3371478811 Synthonix Corporation

4-Pyridinesulfonamide

  • MDL number MFCD12913636
  • Cas Number : 65938-88-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/0086.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/0086.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart