SY3371478813 Synthonix Corporation

1H-Pyrazolo[3,4-b]pyridine-3-carboxylic acid, methyl ester

  • MDL number MFCD13176786
  • Cas Number : 916325-83-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin USA
Product Link https://synthonix.com/0120.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/0120.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart