SY3371478916 Synthonix Corporation

(S)-Pyrrolidine-3-carbonitrile hydrochloride

  • MDL number MFCD11656840
  • Cas Number : 1153950-49-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin USA
Product Link https://synthonix.com/1833.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/1833.html
hstatements H302 , H315 , H318 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart