SY3371478923 Synthonix Corporation

Palladium (II) chloride

  • MDL number MFCD00003558

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Country of Origin China
Product Link https://synthonix.com/2784.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/2784.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart