SY3371478927 Synthonix Corporation

3-Pyrazol-1-yl-benzylamine

  • MDL number MFCD03449005
  • Cas Number : 687635-04-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://synthonix.com/3223.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/3223.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart