SY3371478932 Synthonix Corporation

Pyrrolo[1,2-c]pyrimidine-3-carboxylic acid

  • MDL number MFCD09878579
  • Cas Number : 251102-27-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/4029.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/4029.html
hstatements H315 , H317 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart