SY3371478935 Synthonix Corporation

(S)-1-N-BOC-Piperazine-3-carboxylic acid methyl ester

  • MDL number MFCD04115326
  • Cas Number : 314741-39-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/4430.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/4430.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart