SY3371478943 Synthonix Corporation

Pyrazolo[1,5-a]pyridine-3-carboxylic acid methyl ester

  • MDL number MFCD09752651
  • Cas Number : 63237-84-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/5014.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/5014.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart