SY3371478951 Synthonix Corporation

b-Phthalimidopropionic acid

  • MDL number MFCD00023096
  • Cas Number : 3339-73-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link https://synthonix.com/5090.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/5090.html
hstatements H315 , H317 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart