SY3371478973 Synthonix Corporation

2,4-cis-Piperidine-2,4-dicarboxylic acid

  • MDL number MFCD00057330
  • Cas Number : 84211-45-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/6360.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/6360.html
hstatements H315 , H317 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart