SY3371479052 Synthonix Corporation

N-(Tetrahydro-4-phenyl-2H-pyran-4-yl)acetamide

  • MDL number MFCD21641995
  • Cas Number : 1316852-64-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/0621.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/0621.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart