SY3371479085 Synthonix Corporation

1-(Tert-butoxycarbonyl)-4-hydroxypiperidine-2-carboxylic acid

  • MDL number MFCD01861765
  • Cas Number : 917835-93-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/1242.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/1242.html
hstatements H301 , H315 , H317 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart