SY3371479110 Synthonix Corporation

Triphenylphosphineimine sulfate

  • MDL number MFCD22422287
  • Cas Number : 117672-48-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/1962.html

Safety Information

MSDS https://synthonix.com/1962.html
hstatements H302 ,
Availability details for
Quantity
  • Availability
  • Add to Cart