SY3371479118 Synthonix Corporation

4-(Hydroxymethyl)thiazole

  • MDL number MFCD04115728

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/2165.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/2165.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart