SY3371479135 Synthonix Corporation

4-(Trifluoromethyl)-5-(tributylstannyl)pyrazole

  • MDL number MFCD10699133
  • Cas Number : 790661-62-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin China
Product Link https://synthonix.com/3017.html

Safety Information

pcodes P261 , P273 , P280 ,
MSDS https://synthonix.com/3017.html
hstatements H301 , H302 , H312 , H315 , H318 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart