SY3371479140 Synthonix Corporation

5-Tributylstannyl-1,3-dimethyluracil

  • MDL number MFCD01318970
  • Cas Number : 103408-63-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://synthonix.com/3407.html

Safety Information

pcodes P261 , P273 , P280 ,
MSDS https://synthonix.com/3407.html
hstatements H301 , H312 , H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart