SY3371479156 Synthonix Corporation

4,5,6,7-Tetrahydro-3H-imidazo[4,5-c]pyridine hydrochloride

  • MDL number MFCD09999261
  • Cas Number : 879668-17-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/4238.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/4238.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart