SY3371479158 Synthonix Corporation

5,6,7,8-Tetrahydropyrido[4,3-d]pyrimidine-2,4(1H,3H)-dione hydrochloride

  • MDL number MFCD10699253
  • Cas Number : 1170023-46-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/4274.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/4274.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart