SY3371479160 Synthonix Corporation

7-(Trifluoromethyl)-1,2,3,4-tetrahydroisoquinoline hydrochloride

  • Cas Number : 220247-87-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/4307.html

Safety Information

MSDS https://synthonix.com/4307.html
Availability details for
Quantity
  • Availability
  • Add to Cart