SY3371479185 Synthonix Corporation

5,6,7,8-Tetrahydro-1,6-naphthyridine hydrochloride

  • Cas Number : 1187830-51-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/5331.html

Safety Information

MSDS https://synthonix.com/5331.html
Availability details for
Quantity
  • Availability
  • Add to Cart