SY3371479195 Synthonix Corporation

1-Methyl-1,4,5,6-tetrahydropyrrolo[3,4-c]pyrazole hydrochloride

  • Cas Number : 1187830-68-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/5476.html

Safety Information

MSDS https://synthonix.com/5476.html
Availability details for
Quantity
  • Availability
  • Add to Cart