SY3371479233 Synthonix Corporation

7-Trimethylstannylisoquinoline

  • MDL number MFCD22199151
  • Cas Number : 1032315-16-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/5864.html

Safety Information

pcodes P273 , P280 ,
MSDS https://synthonix.com/5864.html
hstatements H301 , H312 , H315 , H319 ,
Availability details for
Quantity
  • Availability
  • Add to Cart