SY3371479238 Synthonix Corporation

2-Triisopropylsilyloxazole

  • MDL number MFCD17169832
  • Cas Number : 433332-27-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/5929.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/5929.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart