SY3371479241 Synthonix Corporation

2H-Thiete-1,1-dioxide

  • MDL number MFCD13180460
  • Cas Number : 7285-32-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link https://synthonix.com/5975.html

Safety Information

MSDS https://synthonix.com/5975.html
hstatements H301 , H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart