SY3371479271 Synthonix Corporation

3-Thietanemethanamine hydrochloride

  • MDL number MFCD26959157
  • Cas Number : 1523606-32-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/6978.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/6978.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart